Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand:  Ohaus™ 30314910
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15866742
  • £244.00 / Pack of 2
Estimated Shipment: 20-06-2024
to see stock.
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
Specifications
RB
24.5 mm
Rack, 5x25ml D24.5mm RB, 2/pk
2
5 x 25 mL
R-S4x250/4MI Swing Out Rotors
5