Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
£244.00 - £275.00
Specifications
For Use With (Equipment) | R-S4x250/4MI Swing Out Rotors |
---|---|
No. per Pack | 2 |
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
15826752
|
Ohaus™
30314916 |
14 x 5 to 7 mL |
£275.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15836752
|
Ohaus™
30314917 |
10 x 1.6 to 7 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15816752
|
Ohaus™
30314915 |
8 x 10 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15886742
|
Ohaus™
30314912 |
9 x 15 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15896742
|
Ohaus™
30314913 |
7 x 15 mL (17 mm dia. FA) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15806752
|
Ohaus™
30314914 |
7 x 15 mL (17 mm dia. Rim) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15866742
|
Ohaus™
30314910 |
5 x 25 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15876742
|
Ohaus™
30314911 |
3 x 25 mL |
£275.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15856742
|
Ohaus™
30314909 |
1 x 50 mL (29 mm dia. Rim) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15846742
|
Ohaus™
30314908 |
2 x 50 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15816742
|
Ohaus™
30314905 |
3 x 50 mL (29 mm dia. RB) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15826742
|
Ohaus™
30314906 |
3 x 50 mL (29 mm dia. FA) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15836742
|
Ohaus™
30314907 |
1 x 50 mL (34 mm dia. RB) |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15806742
|
Ohaus™
30314904 |
1 x 100 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15896732
|
Ohaus™
30314903 |
1 x 250 mL |
£244.00
Pack of 2 |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
- Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
R-S4x250/4MI Swing Out Rotors | |
2 |