Learn More
Ohaus™ 2 x Centrifuge Rack Adapters, Frontier™ R-S4x250/4MI Swing Out Rotors
Facilitate centrifugation of tubes smaller or greater in number than the original, provided carrier space for Frontier™ R-S4x250/4MI Swing Out Rotors
Brand:  Ohaus™ 30314904
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15806742
  • £244.00 / Pack of 2
In Stock
to see stock.
Description
Description
  • Accommodates centrifuge tubes that are smaller or greater in number than the space provided in the rotor carrier
  • Suitable for use in Frontier™ R-S4x250/4MI Swing Out Rotors
Specifications
Specifications
RB
41 mm
Rack, 1x100ml D41mm RB, 2/pk
2
1 x 100 mL
R-S4x250/4MI Swing Out Rotors
1