Learn More
Fisherbrand™ Two-Way Swivel Scaffolding Connectors
Product Code. 11558112 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
2-Way Connector
Unit Size:
Each
Product Code. Product Type Unit Size
11558112 2-Way Connector Each
1 options
Product Code. 11558112
Estimated Shipment: 07-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Used for fixing two rods or tubes at any angle to each other. Fisherbrand™ Two-Way Swivel Scaffolding Connectors can be tightened by hand.
  • With thumbscrews
  • The two components are restrained from swiveling freely by a tension washer
Specifications
Product Type 2-Way Connector
For Use With (Application) Swivel 2-Way