missing translation for 'onlineSavingsMsg'
Learn More

2-(Trimethylsilylmethyl)allyl acetate, 94%

Product Code. 10340864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10340864 25 g Glass bottle 25g
10260683 1 g Glass bottle 1g
10631115 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10340864 Supplier Thermo Scientific Acros Supplier No. 381380250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 72047-94-0 | C9H18O2Si | 186.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72047-94-0
Molecular Formula C9H18O2Si
Molecular Weight (g/mol) 186.33
MDL Number MFCD00075170
InChI Key IKQWABMHZKGCLX-UHFFFAOYSA-N
Synonym 2-trimethylsilylmethyl allyl acetate, 2-trimethylsilyl methyl allyl acetate, 2-acetoxymethyl-3-trimethylsilyl propene, 2-trimethylsilyl methyl-2-propen-1-yl acetate, 2-trimethylsilyl methyl prop-2-en-1-yl acetate, acmc-1bfax, 2-trimethylsilylmethyl prop-2-enyl acetate, 2-acetoxymethyl allyl-trimethylsilane
PubChem CID 4428347
SMILES CC(=O)OCC(=C)C[Si](C)(C)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8770g/mL
Boiling Point 95°C (7.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2C(=CH2)CH2Si(CH3)3
Refractive Index 1.4380 to 1.44
Quantity 25 g
Specific Gravity 0.877
Solubility Information Solubility in water: reacts
IUPAC Name 2-(trimethylsilylmethyl)prop-2-enyl acetate
Formula Weight 186.23
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Trimethylsilylmethyl)allyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Trimethylsilylmethyl)allyl acetate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.