accessibility menu, dialog, popup

UserName

H3COOH2CSiCH3CH3H3C

CAS RN 72047-94-0

IUPAC Name: 2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate
Synonyms: 2-[(trimethylsilyl)methyl]prop-2-en-1-yl acetate
Molecular Weight (g/mol): 186.33
Molecular Formula: C9H18O2Si
InChi Key: IKQWABMHZKGCLX-UHFFFAOYSA-N
SMILES: CC(=O)OCC(=C)C[Si](C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results