missing translation for 'onlineSavingsMsg'
Learn More

2-tert-Butylpropane-1,3-diol, 98%, Thermo Scientific Chemicals

Product Code. 11353836
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353836 1 g 1g
11363836 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11353836 Supplier Thermo Scientific Alfa Aesar Supplier No. L16927.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2819-05-8
MDL Number MFCD01075745

Specifications

Melting Point 54°C to 56°C
Boiling Point 100°C to 102°C (2 mmHg)
Molecular Formula C7H16O2
Quantity 1 g
Beilstein 1900687
Synonym 2-tert-butyl-1,3-propanediol, 1,3-propanediol, 2-tert-butyl, 1,3-propanediol, 2-1,1-dimethylethyl, acmc-20anmo, 2-t-butylpropane-1,3-diol, 2-tert.-butylpropane-1,3-diol, 2-tert-butyl propane-1,3-diol, 2-tert-butylpropane-1,3-diol, 2-1,1-dimethylethyl propane-1,3-diol, 1,3-propanediol,2-1,1-dimethylethyl
InChI Key LQPVVSDKTCYYBZ-UHFFFAOYSA-N
SMILES CC(C)(C)C(CO)CO
IUPAC Name 2-tert-butylpropane-1,3-diol
Molecular Weight (g/mol) 132.20
PubChem CID 100226
Formula Weight 132.2
Percent Purity 98%
Chemical Name or Material 2-tert-Butylpropane-1,3-diol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.