missing translation for 'onlineSavingsMsg'
Learn More

(2-Pyrimidylthio)acetic acid, 98%, Thermo Scientific™

Product Code. 10693172 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10693172 5 g Glass bottle 5g
10788932 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10693172 Supplier Thermo Scientific Acros Supplier No. 334490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 88768-45-0
Molecular Formula C6H5N2O2S
Molecular Weight (g/mol) 169.18
MDL Number MFCD00010287
InChI Key NIEOYUNNKKAQKI-UHFFFAOYSA-M
Synonym 2-pyrimidylthio acetic acid, pyrimidin-2-ylthio acetic acid, 2-pyrimidin-2-ylthio acetic acid, pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidinylthio acetic acid, acetic acid, 2-pyrimidinylthio, 2-carboxymethylthio pyrimidine, 2-pyrimidin-2-ylsulfanyl acetic acid, 2-pyrimidin-2-ylthioacetic acid, acetic acid, 2-2-pyrimidinylthio
PubChem CID 581022
IUPAC Name 2-pyrimidin-2-ylsulfanylacetic acid
SMILES [O-]C(=O)CSC1=NC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 200°C to 203°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 170.19
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (2-Pyrimidylthio)acetic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (2-Pyrimidylthio)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.