accessibility menu, dialog, popup

UserName

CAS RN 88768-45-0

OOSNN

CAS RN 88768-45-0

IUPAC Name: 2-(pyrimidin-2-ylsulfanyl)acetate
Synonyms: 2-(pyrimidin-2-ylsulfanyl)acetate
Molecular Weight (g/mol): 169.18
Molecular Formula: C6H5N2O2S
InChi Key: NIEOYUNNKKAQKI-UHFFFAOYSA-M
SMILES: [O-]C(=O)CSC1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)