missing translation for 'onlineSavingsMsg'
Learn More

2-Phenylthio-5-propionylphenylacetic Acid, TRC

Product Code. 30418012
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30418012 1 g
30421312 25 g
30415892 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418012 Supplier Toronto Research Chemicals Supplier No. TRCP3370501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103918-73-6
Molecular Formula C17H16O3S
Molecular Weight (g/mol) 300.37
Synonym 5-(1-Oxopropyl)-2-(phenylthio)benzeneacetic Acid
IUPAC Name 2-(2-phenylsulfanyl-5-propanoylphenyl)acetic acid
SMILES CCC(=O)c1ccc(Sc2ccccc2)c(CC(=O)O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H16O3S/c1-2-15(18)12-8-9-16(13(10-12)11-17(19)20)21-14-6-4-3-5-7-14/h3-10H,2,11H2,1H3,(H,19,20)
Formula Weight 300.082
Chemical Name or Material 2-Phenylthio-5-propionylphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.