CAS RN 103918-73-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Phenylthio-5-propionylphenylacetic Acid, TRC
CAS: 103918-73-6 Molecular Formula: C17H16O3S Molecular Weight (g/mol): 300.37 Synonym: 5-(1-Oxopropyl)-2-(phenylthio)benzeneacetic Acid IUPAC Name: 2-(2-phenylsulfanyl-5-propanoylphenyl)acetic acid SMILES: CCC(=O)c1ccc(Sc2ccccc2)c(CC(=O)O)c1