missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybiphenyl, 98+%, Thermo Scientific™

Product Code. 11342406
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11342406 5 g 5g
11352406 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342406 Supplier Thermo Scientific Alfa Aesar Supplier No. L06183.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6738-04-1
MDL Number MFCD00003035

Specifications

Melting Point 47°C to 50°C
Molecular Formula C18H14O
Quantity 5 g
Beilstein 2371408
Synonym 2-phenoxybiphenyl, 2-phenoxy-1,1'-biphenyl, 2-phenyloxybiphenyl, phenoxy-2-diphenyl, 1,1'-biphenyl, phenoxy, 1,1'-biphenyl, 2-phenoxy, o-diphenyl phenyl ether, 2-biphenylyl phenyl ether, ether, 2-biphenylyl phenyl, phenyl biphenyl ether
InChI Key UHJWZORSTYATLW-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=CC=C2OC3=CC=CC=C3
IUPAC Name 1-phenoxy-2-phenylbenzene
Molecular Weight (g/mol) 246.31
PubChem CID 81220
Formula Weight 246.31
Percent Purity ≥98%
Chemical Name or Material 2-Phenoxybiphenyl
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.