accessibility menu, dialog, popup

UserName

CAS RN 6738-04-1

O

CAS RN 6738-04-1

IUPAC Name: 2-phenoxy-1,1'-biphenyl
Synonyms: 2-phenoxy-1,1'-biphenyl
Molecular Weight (g/mol): 246.31
Molecular Formula: C18H14O
InChi Key: UHJWZORSTYATLW-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results