missing translation for 'onlineSavingsMsg'
Learn More

2'-O-Rhamnosylicariside II, MedChemExpress

Product Code. 16409278
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16409278 10 mg 10mg
16401708 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16409278 Supplier Medchem Express Supplier No. HYN2289
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 2'-O-Rhamnosylicariside II is a flavonoid glycoside compound and might be beneficial for improving postmenopausal osteoporosis.

2'-O-Rhamnosylicariside II is a flavonoid glycoside compound and might be beneficial for improving postmenopausal osteoporosis.

Specifications

Chemical Name or Material 2'-O-Rhamnosylicariside II
CAS 135293-13-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H40O14
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (151.36 mM; Need ultrasonic)
SMILES O=C1C(O[C@H]2[C@@H]([C@@H]([C@H]([C@H](C)O2)O)O)O[C@H]3[C@@H]([C@@H]([C@H]([C@H](C)O3)O)O)O)=C(C4=CC=C(OC)C=C4)OC5=C(C/C=C(C)\C)C(O)=CC(O)=C15
Molecular Weight (g/mol) 660.66
Formula Weight 660.66
Percent Purity 98.51%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.