missing translation for 'onlineSavingsMsg'
Learn More

2-Nonyn-1-ol, 96%

Product Code. 11322496
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11322496 5 g 5g
11332496 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11322496 Supplier Thermo Scientific Alfa Aesar Supplier No. L06380.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5921-73-3 | C9H16O | 140.23 g/mol

2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nonyn-1-ol is used as an organic chemical synthesis intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5921-73-3
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00041566
InChI Key XERDOEKKHDALKJ-UHFFFAOYSA-N
Synonym 2-nonyn-1-ol, pubchem13067, acmc-1apv0
PubChem CID 80017
IUPAC Name non-2-yn-1-ol
SMILES CCCCCCC#CCO

Specifications

Density 0.873
Boiling Point 109°C to 111°C (11 mmHg)
Refractive Index 1.456
Quantity 5 g
Beilstein 1745753
Solubility Information Not miscible in water.
Formula Weight 140.23
Percent Purity 96%
Chemical Name or Material 2-Nonyn-1-ol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.