accessibility menu, dialog, popup

UserName

H3COH

CAS RN 5921-73-3

IUPAC Name: non-2-yn-1-ol
Synonyms: non-2-yn-1-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: XERDOEKKHDALKJ-UHFFFAOYSA-N
SMILES: CCCCCCC#CCO
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)