missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl chloride, 98%

Product Code. 10173670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10173670 50 g Glass bottle 50g
10511971 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10173670 Supplier Thermo Scientific Acros Supplier No. 148160500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 612-23-7 | C7H6ClNO2 | 171.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 612-23-7
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00007185
InChI Key BXCBUWKTXLWPSB-UHFFFAOYSA-N
Synonym 2-nitrobenzyl chloride, 1-chloromethyl-2-nitrobenzene, o-nitrobenzyl chloride, alpha-chloro-2-nitrotoluene, benzene, 1-chloromethyl-2-nitro, 2-nitrobenzylchloride, unii-hb8u484npm, alpha-chloro-o-nitrotoluene, ccris 2323, toluene, alpha-chloro-o-nitro
PubChem CID 11921
IUPAC Name 1-(chloromethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CCl)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44°C to 49°C
Color Tan to Yellow
Boiling Point 127°C to 133°C (10.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CH2Cl
Quantity 50 g
Beilstein 05, 327
Solubility Information Solubility in water: insoluble in water
Formula Weight 171.58
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material 2-Nitrobenzyl chloride, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.