accessibility menu, dialog, popup

UserName

ONOCl

CAS RN 612-23-7

IUPAC Name: 1-(chloromethyl)-2-nitrobenzene
Synonyms: 2-nitro-benzylchlorid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: BXCBUWKTXLWPSB-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1CCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)