missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11364106
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11364106 5 g 5g
11354106 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11364106 Supplier Thermo Scientific Alfa Aesar Supplier No. L19238.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 837-95-6
Molecular Formula C7H3ClF3NO4S
Molecular Weight (g/mol) 289.61
MDL Number MFCD00013456
InChI Key KXEMVGQZZLRLBE-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzenesulfonyl chloride, 2-nitro-4-trifluoromethyl benzene-1-sulfonyl chloride, 2-nitro-4-trifluoromethyl benzenesulfonylchloride, pubchem5675, 2-nitro-4-trifluromethylbenzenesulfonylchloride, 2-nitro-4-trifluoromethyl benzene-sulfonyl chlor, 2-nitro-4-trifluoromethyl phenylsulfonyl chloride, 2-nitro-4 trifluoromethyl benzene sulfonyl chloride, 2-nitro-4-trifluoromethyl benzenesulphonyl chloride
PubChem CID 2775780
SMILES [O-][N+](=O)C1=C(C=CC(=C1)C(F)(F)F)S(Cl)(=O)=O

Specifications

Melting Point 42°C to 44°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 2663805
Sensitivity Moisture sensitive
IUPAC Name 2-nitro-4-(trifluoromethyl)benzenesulfonyl chloride
Formula Weight 289.62
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.