accessibility menu, dialog, popup

UserName

FFFNOOSOOCl

CAS RN 837-95-6

IUPAC Name: 2-nitro-4-(trifluoromethyl)benzene-1-sulfonyl chloride
Synonyms: 2-nitro-4-(trifluoromethyl)benzenesulfonyl chloride
Molecular Weight (g/mol): 289.61
Molecular Formula: C7H3ClF3NO4S
InChi Key: KXEMVGQZZLRLBE-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(=C1)C(F)(F)F)S(Cl)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results