missing translation for 'onlineSavingsMsg'
Learn More

2-Methylpropionic-2-d1 Acid, TRC

Product Code. 30391916
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30391916 50 mg -
30400670 10 mg 10mg
30407460 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391916 Supplier Toronto Research Chemicals Supplier No. TRCI78100650MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19136-93-7
Molecular Formula C4 D H7 O2
Molecular Weight (g/mol) 89.1113
Synonym Propanoic-2-d acid, 2-methyl- (9CI, ACI), 2-Methylpropanoic-2-d acid (ACI), Isobutyric-2-d acid (6CI, 8CI), 2-Methylpropanoic Acid-2-D, 2-Methylpropionic-2-d1 Acid, Isobutyric acid-2-d1
IUPAC Name 2-deuterio-2-methylpropanoic acid
SMILES [2H]C(C)(C)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C4H8O2/c1-3(2)4(5)6/h3H,1-2H3,(H,5,6)/i3D
Formula Weight 89.0587
Chemical Name or Material 2-Methylpropionic-2-d1 Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.