CAS RN 19136-93-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-Methylpropionic-2-d1 Acid, TRC
CAS: 19136-93-7 Molecular Formula: C4 D H7 O2 Molecular Weight (g/mol): 89.1113 Synonym: Propanoic-2-d acid, 2-methyl- (9CI, ACI),2-Methylpropanoic-2-d acid (ACI),Isobutyric-2-d acid (6CI, 8CI),2-Methylpropanoic Acid-2-D,2-Methylpropionic-2-d1 Acid,Isobutyric acid-2-d1 IUPAC Name: 2-deuterio-2-methylpropanoic acid SMILES: [2H]C(C)(C)C(=O)O
2
2-Methylpropionic-2-d1 Acid, CDN
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.