missing translation for 'onlineSavingsMsg'
Learn More

(2-Methylbenzyl)triphenylphosphonium chloride, 98+%, Thermo Scientific™

Product Code. 11479572
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479572 10 g 10g
11489572 50 g 50g
11499572 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479572 Supplier Thermo Scientific Alfa Aesar Supplier No. A10719.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63368-36-5
Molecular Formula C26H24ClP
Molecular Weight (g/mol) 402.90
MDL Number MFCD00031577
InChI Key RDHRQYBAQXBTNT-UHFFFAOYSA-M
Synonym 2-methylbenzyl triphenylphosphonium chloride, phosphonium, 2-methylphenyl methyl triphenyl-, chloride, 2-methylbenzyl triphenylphosphoniumchloride, phosphonium, 2-methylphenyl methyl triphenyl-, chloride 1:1, 2-methylphenyl methyl triphenylphosphanium chloride, c26h24p.cl, 2-methylbenzyl triphenyl phosphonium chloride, acmc-20appb
PubChem CID 113182
IUPAC Name (2-methylphenyl)methyl-triphenylphosphanium;chloride
SMILES [Cl-].CC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 270°C to 275°C
Quantity 10 g
Beilstein 4172676
Sensitivity Hygroscopic
Formula Weight 402.91
Percent Purity ≥98%
Chemical Name or Material (2-Methylbenzyl)triphenylphosphonium chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.