accessibility menu, dialog, popup

UserName

ClCH3P

CAS RN 63368-36-5

IUPAC Name: [(2-methylphenyl)methyl]triphenylphosphanium chloride
Synonyms: [(2-methylphenyl)methyl]triphenylphosphanium chloride
Molecular Weight (g/mol): 402.9
Molecular Formula: C26H24ClP
InChi Key: RDHRQYBAQXBTNT-UHFFFAOYSA-M
SMILES: [Cl-].CC1=CC=CC=C1C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results