missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-buten-1-ol, 98%

Product Code. 10613225 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10613225 1 g Glass bottle 1g
11994621 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10613225 Supplier Thermo Scientific Acros Supplier No. 425690010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4516-90-9 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4516-90-9
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
InChI Key NVGOATMUHKIQQG-UHFFFAOYSA-N
Synonym 2-methyl-3-buten-1-ol, 3-buten-1-ol, 2-methyl, 2-methyl-but-3-en-1-ol, 3-buten-1-ol, 2-methyl-, 2s, acmc-20mpc0, acmc-1bn7w, 2-methyl-3-butene-1-ol, 1-hydroxy-2-methyl-3-butene, nvgoatmuhkiqqg-uhfffaoysa
PubChem CID 94292
IUPAC Name 2-methylbut-3-en-1-ol
SMILES CC(CO)C=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 0.8350g/mL
Boiling Point 120.0°C to 121.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(CH3)CH2OH
Refractive Index 1.4260 to 1.4280 (20°C, 589nm)
Quantity 1 g
Specific Gravity 0.835
Formula Weight 86.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-3-buten-1-ol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-3-buten-1-ol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.