CAS RN 4516-90-9
IUPAC Name:
2-methylbut-3-en-1-ol
Synonyms:
2-methylbut-3-en-1-ol
Molecular Weight (g/mol):
86.13
Molecular Formula:
C5H10O
InChi Key:
NVGOATMUHKIQQG-UHFFFAOYNA-N
SMILES:
CC(CO)C=C
1
–
1
of
1
results
1
2-Methyl-3-buten-1-ol, 98%
CAS: 4516-90-9 Molecular Formula: C5H10O Molecular Weight (g/mol): 86.13 InChI Key: NVGOATMUHKIQQG-UHFFFAOYSA-N Synonym: 2-methyl-3-buten-1-ol,3-buten-1-ol, 2-methyl,2-methyl-but-3-en-1-ol,3-buten-1-ol, 2-methyl-, 2s,acmc-20mpc0,acmc-1bn7w,2-methyl-3-butene-1-ol,1-hydroxy-2-methyl-3-butene,nvgoatmuhkiqqg-uhfffaoysa PubChem CID: 94292 IUPAC Name: 2-methylbut-3-en-1-ol SMILES: CC(CO)C=C