missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-naphthylmagnesium bromide, 0.25 M in 2-MeTHF, Thermo Scientific™

Product Code. 15471126
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471126 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471126 Supplier Thermo Scientific Alfa Aesar Supplier No. H54388.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 21450-90-8
Molecular Formula C11H9BrMg
MDL Number MFCD01311486
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive; May form a precipitate
Synonym 2-methyl-1-naphthylmagnesium bromide, frkjgfapmohvam-uhfffaoysa-m, 2-methyl-1-naphthyl magnesium bromide, magnesium,bromo 2-methyl-1-naphthalenyl, magnesium,2-methyl-1h-naphthalen-1-ide,bromide, 2-methyl-1-naphthylmagnesium bromide, 0.25 m in thf, 2-methyl-1-naphthylmagnesium bromide, 0.25 m in 2-methf, 2-methyl-1-naphthylmagnesium bromide 0.25 m in tetrahydrofuran, grignard reagent
InChI Key FRKJGFAPMOHVAM-UHFFFAOYSA-M
SMILES CC1=C([Mg]Br)C2=CC=CC=C2C=C1
IUPAC Name magnesium;2-methyl-1H-naphthalen-1-ide;bromide
Molecular Weight (g/mol) 245.40
PubChem CID 11791449
Formula Weight 245.4
Concentration or Composition (by Analyte or Components) 0.25M in 2-MeTHF
Chemical Name or Material 2-Methyl-1-naphthylmagnesium bromide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.