missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxythiophene, 99%

Product Code. 11498673
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498673 5 g 5g
11488673 1 g 1g
11408683 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498673 Supplier Thermo Scientific Alfa Aesar Supplier No. A10455.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 16839-97-7 | C5H6OS | 114.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16839-97-7
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014526
InChI Key OKEHURCMYKPVFW-UHFFFAOYSA-N
Synonym thiophene, 2-methoxy, 2-methoxy thiophene, 2-methoxy-thiophene, 2-methyloxy thiophene, pubchem12545, acmc-1bwzz, 2-methoxythiophene, ksc183i2f
PubChem CID 85610
IUPAC Name 2-methoxythiophene
SMILES COC1=CC=CS1

Specifications

Density 1.13
Boiling Point 154°C
Flash Point 43°C (109°F)
Refractive Index 1.528
Quantity 5 g
UN Number UN3271
Beilstein 106271
Formula Weight 114.17
Percent Purity 99%
Chemical Name or Material 2-Methoxythiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.