accessibility menu, dialog, popup

UserName

CAS RN 16839-97-7

H3COS

CAS RN 16839-97-7

IUPAC Name: 2-methoxythiophene
Synonyms: p-methoxythiophene
Molecular Weight (g/mol): 114.16
Molecular Formula: C5H6OS
InChi Key: OKEHURCMYKPVFW-UHFFFAOYSA-N
SMILES: COC1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results