missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylbenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15469827
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469827 5 g 5g
15459827 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469827 Supplier Thermo Scientific Alfa Aesar Supplier No. H53059.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Reactant for coupling reactions, Preparation of selective quinazolinyl-phenol inhibitors of CHK1 as potential antitumors and radioprotectants, Rh-catalyzed asymmetric arylation of aldehydes, Stereoselective hydroarylation reactions, Suzuki-Miyaura cross-coupling.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127972-00-3
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.98
MDL Number MFCD02179465
InChI Key CSVKZOZMPSRLTC-UHFFFAOYSA-N
Synonym 2-methoxy-5-methylphenyl boronic acid, 2-methoxy-5-methylbenzeneboronic acid, 2-methoxy-5-methyl-phenyl boronic acid, 2-methoxy-5-methylphenyl boranediol, 5-methyl-2-methoxyphenylboronic acid, boronic acid, 2-methoxy-5-methylphenyl, boronic acid, b-2-methoxy-5-methylphenyl, pubchem23333, acmc-1byqj, 2-methoxy-5-methyl-phenylboronic acid
PubChem CID 2773485
IUPAC Name (2-methoxy-5-methylphenyl)boronic acid
SMILES COC1=CC=C(C)C=C1B(O)O

Specifications

Melting Point 80°C to 84°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 165.99
Percent Purity 95%
Chemical Name or Material 2-Methoxy-5-methylbenzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.