accessibility menu, dialog, popup

UserName

CAS RN 127972-00-3

IUPAC Name: (2-methoxy-5-methylphenyl)boronic acid
Synonyms: 2-methoxy-5-methylphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: CSVKZOZMPSRLTC-UHFFFAOYSA-N
SMILES: COC1=CC=C(C)C=C1B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results