missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 11461290
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461290 1 g 1g
11471290 5 g 5g
11481290 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11461290 Supplier Thermo Scientific Alfa Aesar Supplier No. H26370.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Methoxy-4-(trifluoromethyl)benzoic acid is used as a synthetic building block.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448-36-2
Molecular Formula C9H7F3O3
Molecular Weight (g/mol) 220.147
MDL Number MFCD04115976
InChI Key QWRIRBUKLBWVNF-UHFFFAOYSA-N
Synonym 2-methoxy-4-trifluoromethyl benzoic acid, benzoic acid, 2-methoxy-4-trifluoromethyl, 4-carboxy-3-methoxybenzotrifluoride, 2-carboxy-5-trifluoromethyl anisole, 2-methoxy-4-trifluoromethyl-benzoic acid, 4-trifluoromethyl-2-methoxy-benzoic acid, 4-trifluoromethyl-2-methoxybenzoic acid
PubChem CID 3690873
IUPAC Name 2-methoxy-4-(trifluoromethyl)benzoic acid
SMILES COC1=C(C=CC(=C1)C(F)(F)F)C(=O)O

Specifications

Melting Point 125°C to 130°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 220.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.