CAS RN 448-36-2
IUPAC Name:
2-methoxy-4-(trifluoromethyl)benzoic acid
Synonyms:
2-methoxy-4-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol):
220.15
Molecular Formula:
C9H7F3O3
InChi Key:
QWRIRBUKLBWVNF-UHFFFAOYSA-N
SMILES:
COC1=C(C=CC(=C1)C(F)(F)F)C(O)=O
1
–
1
of
1
results
1
2-Methoxy-4-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™
CAS: 448-36-2 Molecular Formula: C9H7F3O3 Molecular Weight (g/mol): 220.147 MDL Number: MFCD04115976 InChI Key: QWRIRBUKLBWVNF-UHFFFAOYSA-N Synonym: 2-methoxy-4-trifluoromethyl benzoic acid,benzoic acid, 2-methoxy-4-trifluoromethyl,4-carboxy-3-methoxybenzotrifluoride,2-carboxy-5-trifluoromethyl anisole,2-methoxy-4-trifluoromethyl-benzoic acid,4-trifluoromethyl-2-methoxy-benzoic acid,4-trifluoromethyl-2-methoxybenzoic acid PubChem CID: 3690873 IUPAC Name: 2-methoxy-4-(trifluoromethyl)benzoic acid SMILES: COC1=C(C=CC(=C1)C(F)(F)F)C(=O)O