missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-(trifluoromethyl)aniline, 97%, Thermo Scientific™

Product Code. 15404727
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15404727

Brand: Thermo Scientific Alfa Aesar H32045.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 634187-08-9
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD08532489
InChI Key QOMZMKOUJOLVKC-UHFFFAOYSA-N
Synonym 2-methoxy-3-trifluoromethyl aniline, 3-trifluoromethyl-o-anisidine, 3-amino-2-methoxybenzotrifluoride, 2-amino-6-trifluoromethyl anisole, 2-methoxy-3-trifluoromethyl benzenamine, 2-methoxy-3-trifluoromethyl phenylamine
PubChem CID 20112025
IUPAC Name 2-methoxy-3-(trifluoromethyl)aniline
SMILES COC1=C(C=CC=C1N)C(F)(F)F

Specifications

Quantity 1 g
UN Number UN2810
Solubility Information Not miscible or difficult to mix.
Formula Weight 191.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.