accessibility menu, dialog, popup

UserName

CAS RN 634187-08-9

H3COH2NFFF

CAS RN 634187-08-9

IUPAC Name: 2-methoxy-3-(trifluoromethyl)aniline
Synonyms: 2-methoxy-3-(trifluoromethyl)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: QOMZMKOUJOLVKC-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1N)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results