missing translation for 'onlineSavingsMsg'
Learn More

2-Ketoglutaric Acid-13C1, TRC

Product Code. 30136374
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136374 5 mg 5mg
30003210 0.5 mg 0.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136374 Supplier Toronto Research Chemicals Supplier No. TRCK1912025MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108395-15-9
Molecular Formula 13C C4 H6 O5
Molecular Weight (g/mol) 147.09
Synonym Pentanedioic-1-13C acid, 2-oxo- (9CI), 2-Ketoglutaric Acid-13C1, 2-Oxopentanedioic-1-13C acid, α-Ketoglutaric Acid-13C, 2-Oxoglutaric Acid-1-13C, 2-Oxopentanedioic Acid-1-13C
IUPAC Name 2-oxo(1^{13}C)pentanedioic acid
SMILES O[13C](=O)C(=O)CCC(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C5H6O5/c6-3(5(9)10)1-2-4(7)8/h1-2H2,(H,7,8)(H,9,10)/i5+1
Formula Weight 147.0249
Chemical Name or Material 2-Ketoglutaric Acid-13C1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.