CAS RN 108395-15-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Ketoglutaric Acid-13C1, TRC
CAS: 108395-15-9 Molecular Formula: 13C C4 H6 O5 Molecular Weight (g/mol): 147.09 Synonym: Pentanedioic-1-13C acid, 2-oxo- (9CI),2-Ketoglutaric Acid-13C1,2-Oxopentanedioic-1-13C acid,α-Ketoglutaric Acid-13C,2-Oxoglutaric Acid-1-13C,2-Oxopentanedioic Acid-1-13C IUPAC Name: 2-oxo(1^{13}C)pentanedioic acid SMILES: O[13C](=O)C(=O)CCC(=O)O