missing translation for 'onlineSavingsMsg'
Learn More

2-Keto-D-gulonic Acid Methyl Ester, TRC

Product Code. 30394603
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394603 5 mg
30408494 25 mg
30406463 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394603 Supplier Toronto Research Chemicals Supplier No. TRCK1913105MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67776-07-2
Molecular Formula C7 H12 O7
Molecular Weight (g/mol) 208.17
Synonym Methyl D-xylo-hex-2-ulosonate, Methyl D-sorbosonate, D-xylo-2-Hexulosonic acid methyl ester, 2-Keto-gulonic acid methyl ester, Ascorbic Acid Imp. D (EP)
IUPAC Name methyl (3R,4S,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoate
SMILES COC(=O)C(=O)[C@H](O)[C@@H](O)[C@H](O)CO

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C7H12O7/c1-14-7(13)6(12)5(11)4(10)3(9)2-8/h3-5,8-11H,2H2,1H3/t3-,4+,5-/m1/s1
Formula Weight 208.0583
Chemical Name or Material Methyl D-Sorbosonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.