CAS RN 67776-07-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Keto-D-gulonic Acid Methyl Ester, TRC
CAS: 67776-07-2 Molecular Formula: C7 H12 O7 Molecular Weight (g/mol): 208.17 Synonym: Methyl D-xylo-hex-2-ulosonate,Methyl D-sorbosonate,D-xylo-2-Hexulosonic acid methyl ester,2-Keto-gulonic acid methyl ester,Ascorbic Acid Imp. D (EP) IUPAC Name: methyl (3R,4S,5R)-3,4,5,6-tetrahydroxy-2-oxohexanoate SMILES: COC(=O)C(=O)[C@H](O)[C@@H](O)[C@H](O)CO