missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethylene Ethisterone, TRC

Product Code. 30390105
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390105 100 mg -
30406368 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30390105 Supplier Toronto Research Chemicals Supplier No. TRCH946700100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2787-02-2
Molecular Formula C22 H28 O3
Molecular Weight (g/mol) 340.46
IUPAC Name (2E,8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-2-(hydroxymethylidene)-10,13-dimethyl-1,6,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12C\C(=C/O)\C(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(O)C#C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C22H28O3/c1-4-22(25)10-8-18-16-6-5-15-11-19(24)14(13-23)12-20(15,2)17(16)7-9-21(18,22)3/h1,11,13,16-18,23,25H,5-10,12H2,2-3H3/b14-13+/t16-,17+,18+,20+,21+,22-/m1/s1
Formula Weight 340.2038
Percent Purity >95%
Chemical Name or Material 2-Hydroxymethylene Ethisterone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.