CAS RN 2787-02-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxymethylene Ethisterone, TRC
CAS: 2787-02-2 Molecular Formula: C22 H28 O3 Molecular Weight (g/mol): 340.46 IUPAC Name: (2E,8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-2-(hydroxymethylidene)-10,13-dimethyl-1,6,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one SMILES: C[C@]12C\C(=C/O)\C(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(O)C#C