missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-12-crown-4, 97%

Product Code. 11485230
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485230 0.25 g 0.25g
11495230 1 g 1g
11405240 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485230 Supplier Thermo Scientific Alfa Aesar Supplier No. 044509.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75507-26-5 | C9H18O5 | 206.238 g/mol

Ethylene oxide is important or critical to the production of detergents, thickeners, solvents, plastics, and various organic chemicals such as ethylene glycol, ethanolamines, simple and complex glycols, polyglycol ethers and other compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75507-26-5
Molecular Formula C9H18O5
Molecular Weight (g/mol) 206.238
MDL Number MFCD00067524
InChI Key NJIPEIQHUNDGPY-UHFFFAOYSA-N
PubChem CID 2733085
IUPAC Name 1,4,7,10-tetraoxacyclododec-2-ylmethanol
SMILES C1COCCOC(COCCO1)CO

Specifications

Density 1.20 g/mL
Boiling Point 115°C (0.04 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.48
Quantity 0.25 g
Beilstein 4661968
Synonym 1,4,7,10-tetraoxacyclododecane-2-methanol, 2-hydroxymethyl-12-crown-4, 2-hydroxymethyl-12-crown4-ether, 1,4,7,10-tetraoxacyclododecan-2-ylmethanol, 2-hydroxymethyl-12-crown, 2-hydroxymethyl-12-crown 4-ether, 2-hydroxymethyl-12-crown-4-ether, 2-hydroxymethyl-1,2-crown 4-ether, 1,4,7,10-tetraoxacyclododecan-2-methanol, 2,5,8,11-tetraoxacyclododecyl methan-1-ol
Solubility Information Miscible in water. Soluble in non-polar solvents. Insoluble in water.
Formula Weight 206.24
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-12-crown-4
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.