CAS RN 75507-26-5
IUPAC Name:
[(2S)-1,4,7,10-tetraoxacyclododecan-2-yl]methanol
Synonyms:
(2S)-1,4,7,10-tetraoxacyclododecan-2-ylmethanol
Molecular Weight (g/mol):
206.24
Molecular Formula:
C9H18O5
InChi Key:
NJIPEIQHUNDGPY-VIFPVBQESA-N
SMILES:
OC[C@H]1COCCOCCOCCO1
1
–
1
of
1
results
1
2-Hydroxymethyl-12-crown-4, 97%
CAS: 75507-26-5 Molecular Formula: C9H18O5 Molecular Weight (g/mol): 206.238 MDL Number: MFCD00067524 InChI Key: NJIPEIQHUNDGPY-UHFFFAOYSA-N PubChem CID: 2733085 IUPAC Name: 1,4,7,10-tetraoxacyclododec-2-ylmethanol SMILES: C1COCCOC(COCCO1)CO