missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyisoquinoline-1,3(2H,4H)-dione, TRC

Product Code. 30396955
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396955 10 mg 10mg
30400086 100 mg 100mg
30403225 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396955 Supplier Toronto Research Chemicals Supplier No. TRCH93980010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6890-08-0
Molecular Formula C9 H7 N O3
Molecular Weight (g/mol) 177.16
Synonym 2-Hydroxy-1,3(2H,4H)-isoquinolinedione
IUPAC Name 2-hydroxy-4H-isoquinoline-1,3-dione
SMILES ON1C(=O)Cc2ccccc2C1=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H7NO3/c11-8-5-6-3-1-2-4-7(6)9(12)10(8)13/h1-4,13H,5H2
Formula Weight 177.0426
Chemical Name or Material 2-Hydroxyisoquinoline-1,3(2H,4H)-dione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.