CAS RN 6890-08-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxyisoquinoline-1,3(2H,4H)-dione, TRC
CAS: 6890-08-0 Molecular Formula: C9 H7 N O3 Molecular Weight (g/mol): 177.16 Synonym: 2-Hydroxy-1,3(2H,4H)-isoquinolinedione IUPAC Name: 2-hydroxy-4H-isoquinoline-1,3-dione SMILES: ON1C(=O)Cc2ccccc2C1=O