missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyerlotinib, TRC

Product Code. 30401677
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401677 10 mg
30401133 5 mg
30392826 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401677 Supplier Toronto Research Chemicals Supplier No. TRCH10104010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2306363-09-5
Molecular Formula C22H23N3O5
Molecular Weight (g/mol) 409.44
Synonym 4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)-2(1H)-quinazolinone
IUPAC Name 4-(3-ethynylanilino)-6,7-bis(2-methoxyethoxy)-1H-quinazolin-2-one
SMILES COCCOc1cc2NC(=O)N=C(Nc3cccc(c3)C#C)c2cc1OCCOC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C22H23N3O5/c1-4-15-6-5-7-16(12-15)23-21-17-13-19(29-10-8-27-2)20(30-11-9-28-3)14-18(17)24-22(26)25-21/h1,5-7,12-14H,8-11H2,2-3H3,(H2,23,24,25,26)
Formula Weight 409.1638
Chemical Name or Material 2-Hydroxyerlotinib
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.