CAS RN 2306363-09-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxyerlotinib, TRC
CAS: 2306363-09-5 Molecular Formula: C22H23N3O5 Molecular Weight (g/mol): 409.44 Synonym: 4-[(3-Ethynylphenyl)amino]-6,7-bis(2-methoxyethoxy)-2(1H)-quinazolinone IUPAC Name: 4-(3-ethynylanilino)-6,7-bis(2-methoxyethoxy)-1H-quinazolin-2-one SMILES: COCCOc1cc2NC(=O)N=C(Nc3cccc(c3)C#C)c2cc1OCCOC