missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxycyclopent-2-en-1-one, TRC

Product Code. 30392565
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392565 50 mg
30391710 100 mg
30401770 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392565 Supplier Toronto Research Chemicals Supplier No. TRCH95407350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10493-98-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.09
Synonym 2-Hydroxy-Cyclopent-2-en-1-one
IUPAC Name 2-hydroxycyclopent-2-en-1-one
SMILES OC1=CCCC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H6O2/c6-4-2-1-3-5(4)7/h2,6H,1,3H2
Formula Weight 98.0368
Chemical Name or Material 2-Hydroxycyclopent-2-en-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.