CAS RN 10493-98-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Hydroxycyclopent-2-en-1-one, TRC
CAS: 10493-98-8 Molecular Formula: C5H6O2 Molecular Weight (g/mol): 98.09 Synonym: 2-Hydroxy-Cyclopent-2-en-1-one IUPAC Name: 2-hydroxycyclopent-2-en-1-one SMILES: OC1=CCCC1=O