missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy Nabumetone, TRC

Product Code. 30407736
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407736 100 mg -
30394719 50 mg -
30404730 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30407736 Supplier Toronto Research Chemicals Supplier No. TRCH950315100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65726-24-1
Molecular Formula C15 H18 O2
Molecular Weight (g/mol) 230.3
Synonym (2RS)-4-(6-Methoxynaphthalen-2-yl)butan-2-ol, Nabumetone Imp. C (EP)
IUPAC Name 4-(6-methoxynaphthalen-2-yl)butan-2-ol
SMILES COc1ccc2cc(CCC(C)O)ccc2c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C15H18O2/c1-11(16)3-4-12-5-6-14-10-15(17-2)8-7-13(14)9-12/h5-11,16H,3-4H2,1-2H3
Formula Weight 230.1307
Chemical Name or Material (2RS)-4-(6-Methoxynaphthalen-2-yl)butan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.